Anti-LGMN Mouse Monoclonal Antibody [clone: M2]
Catalog # ABNOH00005641-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Legumain
- Antigen symbol:LGMN
- Clonality:Monoclonal
- Clone:M2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:5641
- Antigen synonyms:AEP|LGMN1|PRSC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: LGMN
Clonality: Monoclonal
Clone: M2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human