Anti-DUSP5 Mouse Monoclonal Antibody [clone: 2F3]
Catalog # ABNOH00001847-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Dual specificity phosphatase 5
- Antigen symbol:DUSP5
- Clonality:Monoclonal
- Clone:2F3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1847
- Antigen synonyms:DUSP|HVH3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: DUSP5
Clonality: Monoclonal
Clone: 2F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat