Anti-ATF6B Mouse Monoclonal Antibody [clone: 4D10]
Catalog # ABNOH00001388-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Activating transcription factor 6 beta
- Antigen symbol:ATF6B
- Clonality:Monoclonal
- Clone:4D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1388
- Antigen synonyms:G13|CREB-RP|CREBL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ATF6B
Clonality: Monoclonal
Clone: 4D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human