Anti-CBS Mouse Monoclonal Antibody [clone: 6A9]
Catalog # ABNOH00000875-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cystathionine-beta-synthase
- Antigen symbol:CBS
- Clonality:Monoclonal
- Clone:6A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:875
- Antigen synonyms:cystathionine beta-synthase|beta-thionase|serine sulfhydrase|HIP4|methylcysteine synthase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CBS
Clonality: Monoclonal
Clone: 6A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human