Order Entry
France
ContactUsLinkComponent
Anti-CBS Mouse Monoclonal Antibody [clone: 3D10]
Anti-CBS Mouse Monoclonal Antibody [clone: 3D10]
  ABNOH00000875-M02
 :  Abnova
undefined
Anti-CBS Mouse Monoclonal Antibody [clone: 3D10]
  ABNOH00000875-M02
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Cystathionine-beta-synthase
  • Antigen symbol:
    CBS
  • Clonality:
    Monoclonal
  • Clone:
    3D10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    875
  • Antigen synonyms:
    cystathionine beta-synthase|beta-thionase|serine sulfhydrase|HIP4|methylcysteine synthase
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: CBS
Clonality: Monoclonal
Clone: 3D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human