Order Entry
France
Orders LinkContactUsLinkComponent
Anti-BLM Rabbit Polyclonal Antibody

Anti-BLM Rabbit Polyclonal Antibody

Catalog #
Anti-BLM Rabbit Polyclonal Antibody
Catalog #
Supplier:  
Anti-BLM Rabbit Polyclonal Antibody
Product Details & Documents

Recommended Dilutions:

Type: Primary
Antigen: BLM (Bloom syndrome, RecQ helicase-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Documents