Order Entry
France
ContactUsLinkComponent
Anti-Myc Tag None Monoclonal Antibody
Anti-Myc Tag None Monoclonal Antibody
Catalog # ORIGTA100010
Supplier:  OriGene
Anti-Myc Tag None Monoclonal Antibody
Catalog # ORIGTA100010
Supplier:  OriGene

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    Myc tag
  • Antigen symbol:
    MYC
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Specifications

About this item

Type: Tag
Antigen: Myc Tag (Myc tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity: