Order Entry
France
ContactUsLinkComponent
Anti-ITGAM Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ICRF44]
Anti-ITGAM Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ICRF44]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-ITGAM Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ICRF44]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 50-0118-T025
  • 50-0118-T100
  • Antigen symbol:
  • ITGAM
  • ITGAM
  • Antigen name:
  • Integrin Alpha M
  • Integrin Alpha M
  • Antigen synonyms:
  • CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
  • CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • ICRF44
  • ICRF44
  • Reactivity:
  • Baboon, Pig, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Baboon, Pig, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3684
  • 3684
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Purification:
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • Cool
  • Cool
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together