Order Entry
France
ContactUsLinkComponent
Anti-CD45RO Mouse Monoclonal Antibody (Biotin) [clone: UCHL1]
Anti-CD45RO Mouse Monoclonal Antibody (Biotin) [clone: UCHL1]
Catalog # 30-0457-U100
Supplier:  Tonbo Biosciences
undefined
Anti-CD45RO Mouse Monoclonal Antibody (Biotin) [clone: UCHL1]
Catalog # 30-0457-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein tyrosine phosphatase receptor type C
  • Antigen symbol:
    PTPRC
  • Clonality:
    Monoclonal
  • Clone:
    UCHL1
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Chimpanzee,
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    5788
  • Antigen synonyms:
    CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

The UCHL1 antibody reacts with the human CD45 isoform known as CD45RO, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns. With their broad cell distribution, CD45 isoforms are critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA and CD45RB.

The UCHL1 antibody is widely used as a marker for human CD45RO expression on thymocytes, activated memory T cells, monocytes, macrophages and granulocytes. The antibody is reported to be cross-reactive for Chimpanzee CD45RO.

Type: Primary
Antigen: CD45RO (CD45RO)
Clonality: monoclonal
Clone: UCHL1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: