About this item
The IM7 antibody recognises CD44, a ubiquitously expressed cell surface receptor which is important for extracellular matrix organisation, cell-cell and cell-matrix adhesion and migration. CD44 may be expressed in a number of different isoforms (splice variants) from the most typical or 'standard' form, known as CD44s, to variants designated CD44v, e.g. CD44v1 or CD44v6. These receptors interact with several ligands, but most often associate with an extracellular matrix component hyaluronate, through which it mediates adhesion.
The IM7 antibody may be used for detection of all isoforms of CD44, as it recognises constant epitopes near the extracellular proximal domain. (Xu et al, 2002, J. Leukoc. Biol. 72:1133-1141). It has been reported to be cross-reactive with many non-human species including Baboon, Chimpanzee, Cynomolgus, Rhesus, Horse, Cow, Pig, Dog and Cat CD44.
Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: monoclonal
Clone: IM7
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity:
Specifications
- Catalog No:
- 25-0441-U100
- 25-0441-U025
- Antigen symbol:
- CD44
- CD44
- Antigen name:
- CD44 antigen
- CD44 antigen
- Antigen synonyms:
- HERMES|AW121933|AW146109|AU023126|Pgp-1|Ly-24
- HERMES|AW121933|AW146109|AU023126|Pgp-1|Ly-24
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)/Cy7®
- APC (Allophycocyanin)/Cy7®
- Clone:
- IM7
- IM7
- Reactivity:
- Baboon, Pig, Horse, Cynomolgus monkey, Macaque, Chimpanzee, Human, Dog, Mouse, Cat
- Baboon, Pig, Horse, Cynomolgus monkey, Macaque, Chimpanzee, Human, Dog, Mouse, Cat
- Host:
- Rat
- Rat
- Gene ID:
- 12505
- 12505
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Purification:
- Yes
- Yes
- Cross adsorption:
- No
- No
- Storage temperature:
- Cool
- Cool
- Tested applications:
- Flow Cytometry
- Flow Cytometry