Order Entry
France
ContactUsLinkComponent
 
Anti-ACSS2 Rabbit Polyclonal Antibody
Catalog # ABGEAP9606A
Supplier:  Abgent
undefined
Anti-ACSS2 Rabbit Polyclonal Antibody
Catalog # ABGEAP9606A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Acyl-CoA Synthetase short-chain Family member 2
  • Antigen symbol:
    ACSS2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Center
  • Gene ID:
    80328
  • Antigen synonyms:
    NKG2DL2|N2DL2|ALCAN-alpha|RAET1H
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    EQLRDIQLENYTPKC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,4 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This ULBP2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 88-116 amino acids from the Central region of human ULBP2.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ACSS2 (acyl-CoA synthetase short-chain family member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse