Anti-TIFA Rabbit Polyclonal Antibody
Catalog # ABGEAP9587A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:TRAF-interacting Protein with forkhead-associated Domain
- Antigen symbol:TIFA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Gene ID:26015
- Antigen synonyms:TIFAA|T6BP|T2BP
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:SAFASEPRKRDKLEC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,4 mg/ml
- Shipping temperature:4 °C
- Immunogen:This RPAP1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 282-310 amino acids from the N-terminal region of human RPAP1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TIFA (TRAF-interacting protein with forkhead-associated domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human