Anti-PRKAB2 Rabbit Polyclonal Antibody
Catalog # ABGEAP7046A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase, AMP-activated Non-catalytic Subunit Beta 2
- Antigen symbol:PRKAB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-Terminal (1–100 aa)
- Cross adsorption:No
- Antigen synonyms:BB124140|5730553K21Rik|AW049591
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PRKAB2 (protein kinase, AMP-activated, beta 2 non-catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse