Anti-CBS Rabbit Polyclonal Antibody
ABGEAP6959C
: Abgent
- Antibody type:Primary
- Antigen name:cystathionine-beta-synthase
- Antigen symbol:CBS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:middle
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: CBS (cystathionine-beta-synthase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse