Anti-TBK Rabbit Polyclonal Antibody
Catalog # ABGEAP3627A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:TANK-binding kinase 1
- Antigen symbol:TBK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:TBK1|Serine/threonine-protein kinase TBK1|NAK|NF-kappa-B-activating kinase|TANK-binding kinase 1|T2K
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TBK
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human