Anti-Reptin/TIP49B/RUVB2 Rabbit Polyclonal Antibody
Catalog # ABGEAP1922D
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:RuvB-like 2
- Antigen symbol:RUVBL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:REPTIN|RVB2|TAP54-beta|INO80J|ECP-51|RuvB like AAA ATPase 2|TIP48|TIH2|CGI-46|TIP49B|ECP51
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: Reptin/TIP49B/RUVB2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse