Anti-APP Rabbit Polyclonal Antibody
Catalog # ABGEAP13689C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Amyloid beta (A4) Precursor Protein
- Antigen symbol:APP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:CTFgamma|CVAP|ABETA|APPI|AD1|PN-II|ABPP|AAA|PN2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human