Anti-LARP6 Rabbit Polyclonal Antibody
Catalog # ABGEAP10830A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:La ribonucleoprotein Domain Family member 6
- Antigen symbol:LARP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal (808–821 aa)
- Cross adsorption:No
- Antigen synonyms:ACHN
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: LARP6 (La ribonucleoprotein domain family, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human