Order Entry
France
ContactUsLinkComponent
Anti-TRDMT1 Rabbit Polyclonal Antibody
Anti-TRDMT1 Rabbit Polyclonal Antibody
  ABGEAP1021B
 :  Abgent
undefined
Anti-TRDMT1 Rabbit Polyclonal Antibody
  ABGEAP1021B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    tRNA Aspartic Acid Methyltransferase 1
  • Antigen symbol:
    TRDMT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Center ( 440–455 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    DMNT2|DNMT2|MHSAIIP|PUMET|RNMT1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: TRDMT1 (tRNA aspartic acid methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse