Anti-RPS6KA1 Rabbit Polyclonal Antibody
Catalog # ABGEAP10156A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal Protein S6 Kinase, 90kDa, Polypeptide 1
- Antigen symbol:RPS6KA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:S6K-alpha|MAPKAPK-1c|p90RSK1|MAPK-activated protein kinase 1a|MAPKAPK1C|MAPK-activated protein kinase 1c|MRX19|MAP kinase-activated protein kinase 1c|HU-2|MAP kinase-activated protein kinase 1a|ISPK-1|ribosomal S6 kinase 3|MAP kinase-activated protein kinase 1b|MAPKAP kinase 1c|MAPKAP kinase 1a|r|S6K-alpha2|HU-3|90 kDa ribosomal protein S6 kinase 2|RSK-3|p90-RSK2|ribosomal protein S6 kinase alpha 2|MAPKAPK1A|pp90RSK3|RP11-393H10.3|S6K-alpha 1|RSK1|CLS|S6K-alpha-1|p90RSK2|MAPKAPK-1a|p90-RSK 2|p90-RSK 1|dJ590P13.1 (ribosomal protein S6 kinase|90kD|p90-RSK3|RSK3|HU-1|RSK|90 kDa ribosomal protein S6 kinase 1|p90S6K|ribosomal protein S6 kin|polypeptide 1)|S6K-alpha-2|RSK2|pp90RSK2|MAPKAPK1B|90 kDa ribosomal protein S6 kinase 3|S6K-alpha3|RSK-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: RPS6KA1 (ribosomal protein S6 kinase, 90kDa, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human