Anti-MRPL30 Rabbit Polyclonal Antibody
Catalog # AVIVARP70883_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:mitochondrial ribosomal protein L30
- Antigen symbol:MRPL30
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:51263
- Antigen synonyms:RPML28|MRPL28M|L30MT|MRPL28|L28MT|MRP-L28|MRP-L30
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MRPL30 (mitochondrial ribosomal protein L30)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human