Anti-KRT79 Rabbit Polyclonal Antibody
Catalog # AVIVARP70738_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:keratin 79, type II
- Antigen symbol:KRT79
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:338785
- Antigen synonyms:KRT6L|K6L
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: KRT79 (keratin 79)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human