Anti-OR6C1 Rabbit Polyclonal Antibody
AVIVARP69842_P050
- Antibody type:Primary
- Antigen name:olfactory receptor, family 6, subfamily C, member 1
- Antigen symbol:OR6C1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:390321
- Antigen synonyms:OST267
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: OR6C1 (olfactory receptor, family 6, subfamily C, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human