Anti-AGAP3 Rabbit Polyclonal Antibody
Catalog # AVIVARP69188_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
- Antigen symbol:AGAP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:116988
- Antigen synonyms:CRAG|CENTG3|cnt-g3|AGAP-3|MRIP-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: AGAP3 (ArfGAP with GTPase domain, ankyrin repeat and PH domain 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human