Anti-AAR2 Rabbit Polyclonal Antibody
Catalog # AVIVARP68579_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:AAR2 splicing factor homolog
- Antigen symbol:AAR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:25980
- Antigen synonyms:C20orf4|CGI-23
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: AAR2 (AAR2 splicing factor homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human