Anti-YOD1 Rabbit Polyclonal Antibody
Catalog # AVIVARP67914_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:YOD1 deubiquitinase
- Antigen symbol:YOD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:55432
- Antigen synonyms:PRO0907|OTUD2|DUBA8
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: YOD1 (YOD1 deubiquitinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human