Anti-PPP6R3 Rabbit Polyclonal Antibody
Catalog # AVIVARP62361_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:protein phosphatase 6, regulatory subunit 3
- Antigen symbol:PPP6R3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:55291
- Antigen synonyms:C11orf23|SAP190|SAPS3|SAPLa|PP6R3|SAPL
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PPP6R3 (protein phosphatase 6, regulatory subunit 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: