Anti-EIF3K Rabbit Polyclonal Antibody
Catalog # AVIVARP62169_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 3, subunit K
- Antigen symbol:EIF3K
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:27335
- Antigen synonyms:ARG134, EIF3-p28, EIF3S12, HSPC029, M9, MSTP001, PLAC-24, PLAC24, PRO1474, PTD001
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: EIF3K (eukaryotic translation initiation factor 3, subunit K)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: