Anti-ACKR4 Rabbit Polyclonal Antibody
Catalog # AVIVARP62100_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:atypical chemokine receptor 4
- Antigen symbol:ACKR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:51554
- Antigen synonyms:CCBP2|VSHK1|CCR10|CCX-CKR|CCRL1|CCR11|CKR-11|CCX CKR|CC-CKR-11|PPR1|CCR-11
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ACKR4 (atypical chemokine receptor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Pig, Rat