Anti-H2AFY Rabbit Polyclonal Antibody
Catalog # AVIVARP58283_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:H2A histone family, member Y
- Antigen symbol:H2AFY
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:9555
- Antigen synonyms:H2AFJ|H2AF12M|mH2A1|macroH2A1.2|H2A.y|MACROH2A1.1|H2A/y
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: H2AFY (H2A histone family, member Y)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: