Anti-LHPP Rabbit Polyclonal Antibody
Catalog # AVIVARP57605_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:phospholysine phosphohistidine inorganic pyrophosphate phosphatase
- Antigen symbol:LHPP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:64077
- Antigen synonyms:HDHD2B
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: LHPP (phospholysine phosphohistidine inorganic pyrophosphate phosphatase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: