Anti-MNS1 Rabbit Polyclonal Antibody
AVIVARP57224_P050
- Antibody type:Primary
- Antigen name:NHS-like 1
- Antigen symbol:NHSL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:57224
- Antigen synonyms:C6orf63
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: MNS1 (meiosis-specific nuclear structural 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: