Anti-MPC1 Rabbit Polyclonal Antibody
Catalog # AVIVARP56834_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:mitochondrial pyruvate carrier 1
- Antigen symbol:MPC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:171087
- Antigen synonyms:Brp44l
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MPC1 (mitochondrial pyruvate carrier 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat