Anti-SDE2 Rabbit Polyclonal Antibody
AVIVARP53064_P050
- Antibody type:Primary
- Antigen name:SDE2 telomere maintenance homolog (S. pombe)
- Antigen symbol:SDE2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:163859
- Antigen synonyms:dJ671D7.1|C1orf55
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: SDE2 (SDE2 telomere maintenance homolog (S. pombe))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: