Anti-TC2N Rabbit Polyclonal Antibody
AVIVARP52895_P050
- Antibody type:Primary
- Antigen name:tandem C2 domains, nuclear
- Antigen symbol:TC2N
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:123036
- Antigen synonyms:C2CD1|Tac2-N|C14orf47|MTAC2D1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TC2N (tandem C2 domains, nuclear)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: