Anti-SEPHS1 Rabbit Polyclonal Antibody
AVIVARP48737_T100
- Antibody type:Primary
- Antigen name:Selenophosphate synthetase 1
- Antigen symbol:SEPHS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:22929
- Antigen synonyms:SPS|SPS1|SELD
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: SEPHS1 (selenophosphate synthetase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: