Anti-C2CD2L Rabbit Polyclonal Antibody
AVIVARP47081_P050
- Antibody type:Primary
- Antigen name:C2CD2-like
- Antigen symbol:C2CD2L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:9854
- Antigen synonyms:TMEM24|DLNB23
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: C2CD2L (C2CD2-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat