Anti-UGT1A4 Rabbit Polyclonal Antibody
AVIVARP46829_P050
- Antibody type:Primary
- Antigen name:UDP glucuronosyltransferase 1 family, polypeptide A4
- Antigen symbol:UGT1A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:54657
- Antigen synonyms:UGT1.4|UDPGT 1-4|UGT1A4S|UGT1-04|UGT-1D|HUG-BR2|UGT1D|UDPGT
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: UGT1A4 (UDP glucuronosyltransferase 1 family, polypeptide A4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human