Anti-RNF31 Rabbit Polyclonal Antibody
AVIVARP43241_P050
- Antibody type:Primary
- Antigen name:Ring Finger Protein 31
- Antigen symbol:RNF31
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:55072
- Antigen synonyms:ZIBRA|HOIP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:100 µl
Type: Primary
Antigen: RNF31 (ring finger protein 31)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: