Anti-SF3A1 Rabbit Polyclonal Antibody
AVIVARP40616_P050
- Antibody type:Primary
- Antigen name:splicing factor 3a, subunit 1, 120kDa
- Antigen symbol:SF3A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:10291
- Antigen synonyms:PRP21|PRPF21|SAP114|SF3A120
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: SF3A1 (splicing factor 3a, subunit 1, 120kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: