Anti-A630025C20RIK Rabbit Polyclonal Antibody
Catalog # AVIVARP37513_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:ligand dependent nuclear receptor corepressor
- Antigen symbol:LCOR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:212391
- Antigen synonyms:A630025C20Rik|3110023F06Rik|mKIAA1795|Mlr2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: A630025C20RIK
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: