Anti-EIF4A3 Rabbit Polyclonal Antibody
AVIVARP36421_P050
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 4A3
- Antigen symbol:EIF4A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:9775
- Antigen synonyms:MUK34|eIF4AIII|NUK34|NMP265|RCPS|DDX48
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: EIF4A3 (eukaryotic translation initiation factor 4A3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: