Anti-ZNF79 Rabbit Polyclonal Antibody
Supplier: Aviva Systems Biology
Certificates
About this item
Type: Primary
Antigen: ZNF79 (zinc finger protein 79)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Specifications
- Catalog No:
- AVIVARP35825_P050
- AVIVARP35825_T100
- Antigen symbol:
- ZNF79
- ZNF79
- Antigen name:
- zinc finger protein 79
- zinc finger protein 79
- Antigen synonyms:
- pT7
- pT7
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 7633
- 7633
- Western blot:
- Yes
- Yes
- Epitope:
- N-terminal
- N-terminal
- Cross adsorption:
- none
- none
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together