Anti-GRIN2B Rabbit Polyclonal Antibody
Catalog # AVIVARP35015_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor, ionotropic, N-methyl D-aspartate 2B
- Antigen symbol:GRIN2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:2904
- Antigen synonyms:hNR3|GluN2B|NR2B|EIEE27|MRD6|NMDAR2B
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: GRIN2B (glutamate receptor, ionotropic, N-methyl D-aspartate 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: