Order Entry
France
ContactUsLinkComponent
 
Anti-MAP1A Rabbit Polyclonal Antibody
  BOSSBS-1847R
 :  Bioss
undefined
Anti-MAP1A Rabbit Polyclonal Antibody
  BOSSBS-1847R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Microtubule-associated protein 1A
  • Antigen symbol:
    MAP1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    none
  • Antigen synonyms:
    MAP1a|MTAP-1|6330416M19Rik|MTAP1|MOTH1|AI853608
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: MAP1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat