Pipette controller, Safetypette
ROTHHYT0.1
- Capacity:1 - 100 ml
- Description:Pipette controller, Safetypette
- Pk:1
Pipette controller for glass and plastic pipettes from 1 to 100 ml.
- Manual pipetting aid for simple and precise working with EX and blow-out pipettes
- Excellent for right and left handed users
- Sturdy, lightweight design
- Silicone pipettor holder is autoclavable
: Supplied with 0,45 µm safety filter and different-coloured cones.