Disinfectant Detergent Surface Cleaner, SURFA' SAFE Premium
Supplier: Laboratoires Anios
Certificates
About this item
Disinfecting detergent foam for walls floors and non invasive medical equipment.
- Simultaneous action: cleaning and disinfection
- Alcohol- and perfume- and CMR-free
- Contains didecyldimethylammonium chloride, excipients
- Quick drying, does not leave a trace
- Activity on enveloped viruses (Rotavirus, PRV, HSV, vaccinia, BVDV)
Certifications: Bactericide EN 1040, EN 13727; Fungicide EN 1275, EN 13624, EN 13697; EN 14562; Virucide 14476
Caution: Use biocides safely. Always read the label and product information before use.
Frequently Bought Together