Visual colour kit, free chlorine
Supplier: Hach
Certificates
About this item
Simple, economical, rapid visual colour determination.
DPD visual test kit method for free chlorine, which can be used to accurately measure the free chlorine levels in the field or in the lab.
Delivery: Kit includes one reagent bottle, stopper, colour comparator box, colour disc, tube, instruction sheet, and a carrying case.
Frequently Bought Together