

Zinc acetate dihydrate 99.0-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:5970-45-6
:
Formula:(H₃CCOO)₂Zn·2H₂O
MDL Number:MFCD00066961
:219.51 g/mol
Merck Index:12,10256
Melting Pt:237 °C
Density:1.74 g/cm³
Storage Temperature:Ambient
EINECS:209-170-2
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Meets ACS Reagent Chemical Requirements:99.0 - 101.0%
- For Laboratory,Research,or Manufacturing Use:≤0.005%
- ACS - Assay ((CH₃COO)₂Zn·2H₂O):99.0 - 101.0 %
- ACS - Insoluble Matter:≤0.005 %
- ACS - pH of 5% Solution at 25°C:6.0 - 7.0
- ACS - Sulfate (SO₄):≤0.002 %
- ACS - Calcium (Ca):≤0.005 %
- ACS - Lead (Pb):≤0.002 %
- ACS - Magnesium (Mg):≤0.005 %
- ACS - Potassium (K):≤0.01 %
- ACS - Sodium (Na):≤0.05 %
- Chloride (Cl):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
Specification Test Results
Related Information
Frequently Bought Together