Order Entry
France
ContactUsLinkComponent
Pipette controller, M-Sarpette®
Pipette controller, M-Sarpette®
Catalog # SARS90.1427
Supplier:  SARSTEDT
undefined
Pipette controller, M-Sarpette®
Catalog # SARS90.1427
Supplier:  SARSTEDT
Specifications
  • Description:
    Manual pipette aid, M-Sarpette®
  • Pk:
    1
Specifications
About this item
This manual bulb pipettor enables easy filling of pipettes. Compactible with pipettes of 1 to 100 ml volume.
  • Ergonomically designed thumb lever controls asprirating and dispensing
  • Discharge button allows for blow-out of residual liquid
  • Manual operation: No batteries or power cord necessary